Learn More
2,3-Dimethoxytoluene, 98+%, Thermo Scientific Chemicals
Catalog Number p-7081552
314.79 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4463-33-6
C9H12O2
152.193
MFCD00008379
WMXFNCKPYCAIQW-UHFFFAOYSA-N
2,3-dimethoxytoluene, 3-methylveratrole, benzene, 1,2-dimethoxy-3-methyl, dimethoxytoluene, 1,2-dimethoxy-3-methyl-benzene, acmc-1ae12, 1-methyl-2,3-dimethoxybenzene, 3-methylcatechol, dimethyl ether, #
78215
1,2-dimethoxy-3-methylbenzene
CC1=C(C(=CC=C1)OC)OC
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4463-33-6
C9H12O2
152.193
MFCD00008379
WMXFNCKPYCAIQW-UHFFFAOYSA-N
2,3-dimethoxytoluene, 3-methylveratrole, benzene, 1,2-dimethoxy-3-methyl, dimethoxytoluene, 1,2-dimethoxy-3-methyl-benzene, acmc-1ae12, 1-methyl-2,3-dimethoxybenzene, 3-methylcatechol, dimethyl ether, #
78215
1,2-dimethoxy-3-methylbenzene
CC1=C(C(=CC=C1)OC)OC
2,3-Dimethoxytoluene has been used to prepare 6,7-dimethoxy-4-hydroxyisochroman-3-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Dimethoxytoluene has been used to prepare 6,7-dimethoxy-4-hydroxyisochroman-3-one.
Solubility
Soluble in Dichloromethane. Not miscible in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.024
Boiling Point 202°C to 203°C
Flash Point 85°C (185°F)
Odor Aromatic
Refractive Index 1.514
Quantity 5 g
Beilstein 1940962
Solubility Information Soluble in Dichloromethane. Not miscible in water.
Formula Weight 152.19
Percent Purity ≥98%
Chemical Name or Material 2,3-Dimethoxytoluene