missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-2,3-diphenylbutane, 95%, Thermo Scientific Chemicals

Catalog Number p-117216
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1889-67-4
C18H22
238.37
MFCD00053713
HGTUJZTUQFXBIH-UHFFFAOYSA-N
2,3-dimethyl-2,3-diphenylbutane, dicumene, 2,3-dimethylbutane-2,3-diyl dibenzene, dicumyl, butane, 2,3-dimethyl-2,3-diphenyl, benzene, 1,1'-1,1,2,2-tetramethyl-1,2-ethanediyl bis, .alpha.,.alpha.'-dicumyl, 2,3-dimethyl-3-phenylbutan-2-yl benzene, bibenzyl, .alpha.,.alpha.,.alpha.',.alpha.'-tetramethyl, 1,1,2-trimethyl-2-phenylpropyl benzene
74681
(2,3-dimethyl-3-phenylbutan-2-yl)benzene
CC(C)(C1=CC=CC=C1)C(C)(C)C2=CC=CC=C2
This item is not returnable. View return policy
1889-67-4
C18H22
238.37
MFCD00053713
HGTUJZTUQFXBIH-UHFFFAOYSA-N
2,3-dimethyl-2,3-diphenylbutane, dicumene, 2,3-dimethylbutane-2,3-diyl dibenzene, dicumyl, butane, 2,3-dimethyl-2,3-diphenyl, benzene, 1,1'-1,1,2,2-tetramethyl-1,2-ethanediyl bis, .alpha.,.alpha.'-dicumyl, 2,3-dimethyl-3-phenylbutan-2-yl benzene, bibenzyl, .alpha.,.alpha.,.alpha.',.alpha.'-tetramethyl, 1,1,2-trimethyl-2-phenylpropyl benzene
74681
(2,3-dimethyl-3-phenylbutan-2-yl)benzene
CC(C)(C1=CC=CC=C1)C(C)(C)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 90°C to 110°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min
Packaging Glass bottle
Linear Formula C6H5C(CH3)2C(CH3)2C6H5
Quantity 25 g
Beilstein 05,396
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in toluene
Formula Weight 238.37
Percent Purity 95%
Physical Form Crystalline Powder, Flakes, or Chunks
Chemical Name or Material 2, 3-Dimethyl-2, 3-diphenylbutane, 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'