Läs mer
(2,3-Dimethylphenyl)-1H-imidazol-5-ylmethanone, TRC
Produktkod. 30008754
Change view
Kvantitet:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008754 1 g
30008235 50 mg
30012587 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008754 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4807801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 91874-85-0
Molekylformel C12H12N2O
Molekylvikt (g/mol) 200.24
Synonym (2,3-dimethylphenyl)-1H-imidazol-4-ylmethanone
LEDER Cc1cccc(C(=O)c2cnc[nH]2)c1C
Specifikationer
Kemiskt namn eller material (2,3-Dimethylphenyl)-1H-imidazol-5-ylmethanone
Kvantitet 1 g
InChI formel InChI=1S/C12H12N2O/c1-8-4-3-5-10(9(8)2)12(15)11-6-13-7-14-11/h3-7H,1-2H3,(H,13,14)
Formel vikt 200.095