Läs mer
2-(3-Ethylureido)-2-oxoacetic Acid, TRC
Produktkod. 30373684
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30373684 50 mg
1 options
Produktkod. 30373684 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE81826050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(3-Ethylureido)-2-oxoacetic Acid
CAS 105919-00-4
Molekylformel C5H8N2O4
Kvantitet 50 mg
Synonym [[(Ethylamino)carbonyl]amino]oxo-acetic Acid, 2-[[(Ethylamino)carbonyl]amino]-2-oxoacetic Acid
InChI formel InChI=1S/C5H8N2O4/c1-2-6-5(11)7-3(8)4(9)10/h2H2,1H3,(H,9,10)(H2,6,7,8,11)
LEDER CCNC(NC(C(O)=O)=O)=O
Molekylvikt (g/mol) 160.13
Formel vikt 160.0484