Läs mer
[2-(3-Methoxyphenyl)cyclohex-1-enyl]-N,N-dimethylmethanamine Hydrochloride, Mikromol™
Produktkod. 18040720
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18040720 25 mg
18082570 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18040720 Leverantör Mikromol Leverantörsnummer MM0007.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
[2-(3-Methoxyphenyl)cyclohex-1-enyl]-N,N-dimethylmethanamine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2-(3-Methoxyphenyl)cyclohex-1-enyl)-N,N-dimethylmethanamine Hydrochloride
CAS 66170-32-9
Analyt- eller komponentnamn (2-(3-Methoxyphenyl)cyclohex-1-enyl)-N,N-dimethylmethanamine Hydrochloride
Frakt skick Room Temperature
Molekylformel C16 H23 N O . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Tramadol Hydrochloride Imp. B (EP), Tramadol Imp. B (EP), [2-(3-Methoxyphenyl)cyclohex-1-enyl]-N,N-dimethylmethanamine Hydrochloride, Tramadol Hydrochloride Imp. B (EP) as Hydrochloride, Tramadol Hydrochloride Impurity B as Hydrochloride, Tramadol Impurity B as Hydrochloride
InChI formel InChI=1S/C16H23NO.ClH/c1-17(2)12-14-7-4-5-10-16(14)13-8-6-9-15(11-13)18-3;/h6,8-9,11H,4-5,7,10,12H2,1-3H3;1H
LEDER Cl.COc1cccc(c1)C2=C(CN(C)C)CCCC2
IUPAC-namn 1-[2-(3-methoxyphenyl)cyclohexen-1-yl]-N,N-dimethylmethanamine;hydrochloride
Molekylvikt (g/mol) 281.82
Kvantitet 25 mg
Formel vikt 281.1546 g/mol