Learn More
Alfa Aesar™ 2-(3-Methoxyphenyl)thiazolidine-4-carboxylic acid, 97%
Brand:  Alfa Aesar™ H62853.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15477916
  • 2256.11 SEK / 1g
Estimated Shipment date
from Supplier 01-02-2018
Add to basket
Specifications
Chemical Identifiers
2-(3-Methoxyphenyl)thiazolidine-4-carboxylic acid
239.29
MFCD02112793
Specifications
2-(3-Methoxyphenyl)thiazolidine-4-carboxylic acid
97%
MFCD02112793
Solid
239.29
1g