Learn More
2-[3-(Methylamino)-1-(2-thienyl)propyl]-1-naphthalenol Hydrochloride, TRC
Product Code. 30137030
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137030 50 mg 50mg
30006793 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137030 Supplier Toronto Research Chemicals Supplier No. TRCM28721050MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1033719-36-6
Molecular Formula C18H19NOS • HCl
Molecular Weight (g/mol) 297.423646
Synonym Duloxetine EP Impurity E
SMILES CNCCC(C1=CC=CS1)C2=C(O)C3=CC=CC=C3C=C2.[HCl]
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C18H19NOS.ClH/c1-19-11-10-15(17-7-4-12-21-17)16-9-8-13-5-2-3-6-14(13)18(16)20;/h2-9,12,15,19-20H,10-11H2,1H3;1H
Formula Weight 333.0954
Chemical Name or Material 2-[3-(Methylamino)-1-(2-thienyl)propyl]-1-naphthalenol Hydrochloride