Learn More
2,3-Naphthalenediol, Sigma-Aldrich™
Specifications
Specifications
| CAS | 92-44-4 |
| Melting Point | 161°C to 164°C |
| MDL Number | MFCD00004073 |
| Quantity | 50 g |
| Synonym | 2,3-Naphthalenediol, 2,3-Dihydroxynaphthalene, 2,3-Naphthalenediol |
| Molecular Weight (g/mol) | 160.17 |
| Grade | Research |