Läs mer
2'/3'-O-Acetyl ADP Ribose, Disodium Salt (A mixture of 2' and 3'-O-Acetyl ADP Ribose), TRC
Produktkod. 30133484
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30133484 25 mg -
30008834 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133484 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA17380525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1312013-29-8
Molekylformel C17H23N5Na2O15P2
Molekylvikt (g/mol) 645.32
Synonym Adenosine 5'-(trihydrogen diphosphate) P'→5-Ester with D-ribose 2-Acetate Sodium Salt (1:2)
IUPAC-namn tetrasodium;[[(2R,3S,4R)-3-acetyloxy-4,5-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;[[(2R,3R,4R)-4-acetyloxy-3,5-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
LEDER O[C@@H]1[C@H](OC(C)=O)[C@@H](COP(OP(OC[C@H]2O[C@@H](N3C=NC4=C3N=CN=C4N)[C@H](O)[C@@H]2O)(O[Na])=O)(O[Na])=O)OC1O.O[C@H]5[C@@H](OC(C)=O)C(O)O[C@@H]5COP(OP(OC[C@H]6O[C@@H](N7C=NC8=C7N=CN=C8N)[C@H](O)[C@@H]6O)(O[Na])=O)(O[Na])=O
Specifikationer
Kemiskt namn eller material 2'/3'-O-Acetyl ADP Ribose Disodium Salt (A mixture of 2' and 3'-O-Acetyl ADP Ribose)
Kvantitet 25 mg
InChI formel InChI=1S/2C17H25N5O15P2.4Na/c1-6(23)34-13-8(36-17(27)12(13)26)3-33-39(30,31)37-38(28,29)32-2-7-10(24)11(25)16(35-7)22-5-21-9-14(18)19-4-20-15(9)22;1-6(23)34-13-11(25)8(36-17(13)27)3-33-39(30,31)37-38(28,29)32-2-7-10(24)12(26)16(35-7)22-5-21-9-14(18)19-4-20-15(9)22;;;;/h2*4-5,7-8,10-13,16-17,24-27H,2-3H2,1H3,(H,28,29)(H,30,31)(H2,18,19,20);;;;/q;;4*+1/p-4/t2*7-,8-,10-,11-,12-,13-,16-,17?;;;;/m11..../s1
Formel vikt 645.0461