missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Pyridyl)-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2696157
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
39067-29-3
C9H6N2O2S
206.219
MFCD00052304
FOQFGMAZUTUELM-UHFFFAOYSA-N
2-pyridin-3-yl thiazole-4-carboxylic acid, 2-3-pyridyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-1,3-thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 2-3-pyridinyl, 2-pyridin-3-yl-thiazole-4-carboxylic acid, 2-3-pyridyl thiazole-4-carboxylic acid, 2-3-pyridinyl-4-thiazolecarboxylic acid, 2-pyrid-3-yl thiazole-4-carboxylic acid, 2-3-pyridinyl-1,3-thiazole-4-carboxylic acid
736513
2-pyridin-3-yl-1,3-thiazole-4-carboxylic acid
C1=CC(=CN=C1)C2=NC(=CS2)C(=O)O
This item is not returnable. View return policy
39067-29-3
C9H6N2O2S
206.219
MFCD00052304
FOQFGMAZUTUELM-UHFFFAOYSA-N
2-pyridin-3-yl thiazole-4-carboxylic acid, 2-3-pyridyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-3-yl-1,3-thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 2-3-pyridinyl, 2-pyridin-3-yl-thiazole-4-carboxylic acid, 2-3-pyridyl thiazole-4-carboxylic acid, 2-3-pyridinyl-4-thiazolecarboxylic acid, 2-pyrid-3-yl thiazole-4-carboxylic acid, 2-3-pyridinyl-1,3-thiazole-4-carboxylic acid
736513
2-pyridin-3-yl-1,3-thiazole-4-carboxylic acid
C1=CC(=CN=C1)C2=NC(=CS2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 206.22
Percent Purity 97%
Chemical Name or Material 2-(3-Pyridyl)-1, 3-thiazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'