Läs mer
2-(3-Pyridyl)-2-oxazoline, TRC
Produktkod. 30003769
Change view
Kvantitet:
250 mg
500 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003769 500 mg
30003575 250 mg
30012689 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003769 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP995715500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 40055-37-6
Molekylformel C8 H8 N2 O
Molekylvikt (g/mol) 148.16
Synonym 3-(4,5-Dihydrooxazol-2-yl)pyridine, 3-(2-Oxazolin-2-yl)pyridine;
IUPAC-namn 2-pyridin-3-yl-4,5-dihydro-1,3-oxazole
LEDER C1CN=C(O1)c2cccnc2
Specifikationer
Kemiskt namn eller material 2-(3-Pyridyl)-2-oxazoline
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C8H8N2O/c1-2-7(6-9-3-1)8-10-4-5-11-8/h1-3,6H,4-5H2
Formel vikt 148.0637
Procent renhet >95%