Läs mer
2-[3-(R)-[3-(2-(7-Chloro-2-quinolinyl)ethenyl)phenyl]-3-hydroxypropyl]benzoic Acid Methyl Ester, TRC
Produktkod. 30356707
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30356707 100 mg
1 options
Produktkod. 30356707 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC379991100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[3-(R)-[3-(2-(7-Chloro-2-quinolinyl)ethenyl)phenyl]-3-hydroxypropyl]benzoic Acid Methyl Ester
CAS 150026-72-5
Molekylformel C28 H24 Cl N O3
Kvantitet 100 mg
Synonym 2-[(3R)-3-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-hydroxypropyl]benzoic Acid Methyl Ester, [R-(E)]-2-[3-[3-[2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-hydroxypropyl]benzoic Acid Methyl Ester
InChI formel InChI=1S/C28H24ClNO3/c1-33-28(32)25-8-3-2-6-20(25)12-16-27(31)22-7-4-5-19(17-22)9-14-24-15-11-21-10-13-23(29)18-26(21)30-24/h2-11,13-15,17-18,27,31H,12,16H2,1H3/b14-9+/t27-/m1/s1
LEDER COC(=O)c1ccccc1CC[C@@H](O)c2cccc(\C=C\c3ccc4ccc(Cl)cc4n3)c2
IUPAC-namn methyl 2-[(3R)-3-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-hydroxypropyl]benzoate
Molekylvikt (g/mol) 457.948
Formel vikt 457.144