Läs mer
2-[3-(R)-[3-(2-(7-Chloro-2-quinolinyl)ethenyl)phenyl]-3-hydroxypropyl]phenyl-2-propanol, TRC
Produktkod. 30353443
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353443 10 mg
30351994 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353443 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37999610MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 150026-75-8
Molekylformel C29 H28 Cl N O2
Molekylvikt (g/mol) 457.99
Synonym α-[3-[(1E)-2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-2-[(1R)-1-hydroxy-1-methylethyl]benzenepropanol, Benzenepropanol, α-[3-[2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-2-(1-hydroxy-1-methylethyl)-, [R-(E)]- (9CI)
IUPAC-namn (1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propan-1-ol
LEDER CC(C)(O)c1ccccc1CC[C@@H](O)c2cccc(\C=C\c3ccc4ccc(Cl)cc4n3)c2
Specifikationer
Kemiskt namn eller material (1R)-1-(3-((E)-2-(7-Chloro-2-quinolyl)vinyl)phenyl)-3-(2-(1-hydroxy-1-methylethyl)phenyl)propan-1-ol
Kvantitet 10 mg
InChI formel InChI=1S/C29H28ClNO2/c1-29(2,33)26-9-4-3-7-21(26)13-17-28(32)23-8-5-6-20(18-23)10-15-25-16-12-22-11-14-24(30)19-27(22)31-25/h3-12,14-16,18-19,28,32-33H,13,17H2,1-2H3/b15-10+/t28-/m1/s1
Formel vikt 457.1809