Läs mer
2-[3-(S)-[3-(2-(7-Chloro-2-quinolinyl)ethenyl)phenyl]-3-hydroxypropyl]benzoic Acid Methyl Ester, TRC
Produktkod. 30350856
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30350856 10 g
1 options
Produktkod. 30350856 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37999010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[3-(S)-[3-(2-(7-Chloro-2-quinolinyl)ethenyl)phenyl]-3-hydroxypropyl]benzoic acid methyl ester
CAS 142569-69-5
Molekylformel C28 H24 Cl N O3
Kvantitet 10 g
Synonym 2-[3-(S)-[3-(2-(7-Chloro-2-quinolinyl)(E/Z)ethenyl)phenyl]-3-hydroxypropyl]benzoic acid methyl ester
InChI formel InChI=1S/C28H24ClNO3/c1-33-28(32)25-8-3-2-6-20(25)12-16-27(31)22-7-4-5-19(17-22)9-14-24-15-11-21-10-13-23(29)18-26(21)30-24/h2-11,13-15,17-18,27,31H,12,16H2,1H3/t27-/m0/s1
LEDER COC(=O)c1ccccc1CC[C@H](O)c2cccc(C=Cc3ccc4ccc(Cl)cc4n3)c2
IUPAC-namn methyl 2-[(3S)-3-[3-[2-(7-chloro-2-quinolyl)vinyl]phenyl]-3-hydroxy-propyl]benzoate
Molekylvikt (g/mol) 457.95
Formel vikt 457.1445