Läs mer
2-[[4-[[2-(1,1-Dimethylethoxy)phenyl]amino]-6-(1-pyrrolidinyl)-1,3,5-triazin-2-yl]amino]ethanol, TRC
Produktkod. 30378398
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378398 100 mg
30377251 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30378398 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD472965100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 958445-02-8
Molekylformel C19H28N6O2
Molekylvikt (g/mol) 372.46
Synonym 2-[[4-[2-[(2-Methylpropan-2-yl)oxy]anilino]-6-pyrrolidin-1-yl-,1,3,5-triazin-2-yl]amino]ethanol, CID-17757274
LEDER CC(C)(C)Oc1ccccc1Nc2nc(NCCO)nc(n2)N3CCCC3
Specifikationer
Kemiskt namn eller material 2-((4-((2-(1,1-Dimethylethoxy)phenyl)amino)-6-(1-pyrrolidinyl)-1,3,5-triazin-2-yl)amino)ethanol
Kvantitet 100 mg
InChI formel InChI=1S/C19H28N6O2/c1-19(2,3)27-15-9-5-4-8-14(15)21-17-22-16(20-10-13-26)23-18(24-17)25-11-6-7-12-25/h4-5,8-9,26H,6-7,10-13H2,1-3H3,(H2,20,21,22,23,24)
Formel vikt 372.2274