Läs mer
2-[4-[2-(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethanol, TRC
Produktkod. 30367850
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367850 100 mg
30363188 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367850 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC378700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391053-56-7
Molekylformel C19H23ClN2O
Molekylvikt (g/mol) 330.85
Synonym 4-[(4′-Chloro[1,1′-biphenyl]-2-yl)methyl]-1-piperazineethanol (ACI)
IUPAC-namn 2-(4-((4'-chloro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)ethan-1-ol
LEDER ClC(C=C1)=CC=C1C2=C(CN3CCN(CCO)CC3)C=CC=C2
Specifikationer
Kemiskt namn eller material 2-[4-[2-(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethanol
Kvantitet 100 mg
InChI formel InChI=1S/C19H23ClN2O/c20-18-7-5-16(6-8-18)19-4-2-1-3-17(19)15-22-11-9-21(10-12-22)13-14-23/h1-8,23H,9-15H2
Formel vikt 330.15