Läs mer
2-[4-(2,4-Dichloro-5-hydroxyphenoxy)phenoxy]propanoic Acid Methyl Ester, TRC
Produktkod. 30369284
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30369284 25 mg
1 options
Produktkod. 30369284 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43467525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diclofop-methyl-5-hydroxy
CAS 124992-48-9
Molekylformel C16 H14 Cl2 O5
Kvantitet 25 mg
Synonym 2-[4-(2,4-Dichloro-5-hydroxyphenoxy)phenoxy]propanoic Acid Methyl Ester, Propanoic acid, 2-[4-(2,4-dichloro-5-hydroxyphenoxy)phenoxy]-, methyl ester
InChI formel InChI=1S/C16H14Cl2O5/c1-9(16(20)21-2)22-10-3-5-11(6-4-10)23-15-8-14(19)12(17)7-13(15)18/h3-9,19H,1-2H3
LEDER COC(=O)C(C)Oc1ccc(Oc2cc(O)c(Cl)cc2Cl)cc1
IUPAC-namn methyl 2-[4-(2,4-dichloro-5-hydroxyphenoxy)phenoxy]propanoate
Molekylvikt (g/mol) 357.19
Formel vikt 356.0218