Läs mer
2,4,2',4'-Tetrabromodiphenyl Ether, TRC
Produktkod. 17959389
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17959389 2.5 g 2.5g
17938529 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17959389 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2911452.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5436-43-1
Molekylformel C12 H6 Br4 O
Molekylvikt (g/mol) 485.79
Synonym Benzene, 2,4-dibromo-1-(2,4-dibromophenoxy)-, Benzene, 1,1'-oxybis[2,4-dibromo- (9CI), 2,4-Dibromo-1-(2,4-dibromophenoxy)benzene, 2,2',4,4'-Tetrabromodiphenyl ether, 2,2',4,4'-Tetrabromodiphenyl oxide, 2,4,2',4'-Tetrabromodiphenyl ether, BDE 47, NSC 21724, PBDE 47
IUPAC-namn 2,4-dibromo-1-(2,4-dibromophenoxy)benzene
LEDER Brc1ccc(Oc2ccc(Br)cc2Br)c(Br)c1
Specifikationer
Kemiskt namn eller material PBDE 47 (2,2',4,4'-Tetrabromodiphenyl Ether)
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C12H6Br4O/c13-7-1-3-11(9(15)5-7)17-12-4-2-8(14)6-10(12)16/h1-6H
Formel vikt 481.7152
Procent renhet >95%