missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetic Acid, Mikromol™
Beskrivning
2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetic Acid
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetic Acid |
| CAS | 56392-14-4 |
| Analyt- eller komponentnamn | 2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetic Acid |
| Frakt skick | Room Temperature |
| Molekylformel | C14 H21 N O4 |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | 2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetic Acid, Atenolol Imp. G (EP), Atenolol Related Compound C, Atenolol Impurity G |
| InChI formel | InChI=1S/C14H21NO4/c1-10(2)15-8-12(16)9-19-13-5-3-11(4-6-13)7-14(17)18/h3-6,10,12,15-16H,7-9H2,1-2H3,(H,17,18) |
| LEDER | CC(C)NCC(O)COc1ccc(CC(=O)O)cc1 |
| IUPAC-namn | 2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetic acid |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?