missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(3-Chloro-2-hydroxypropoxy)phenyl]acetonitrile, Mikromol™

Product Code. 30049820
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30049820 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049820 Supplier Mikromol Supplier No. MM0013.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[4-(3-Chloro-2-hydroxypropoxy)phenyl]acetonitrile
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-[4-(3-Chloro-2-hydroxypropoxy)phenyl]acetonitrile
CAS 142037-04-5
Analyt- eller komponentnamn 2-[4-(3-Chloro-2-hydroxypropoxy)phenyl]acetonitrile
Frakt skick Ice pack (-20°C)
Molekylformel C11 H12 Cl N O2
Certifikat och efterlevnad ISO 9001
Synonym 4-(3-Chloro-2-hydroxypropoxy)-benzeneacetonitrile, (+/-)-4-(3-chloro-2-hydroxypropoxy)-benzeneacetonitrile
InChI formel InChI=1S/C11H12ClNO2/c12-7-10(14)8-15-11-3-1-9(2-4-11)5-6-13/h1-4,10,14H,5,7-8H2
LEDER OC(CCl)COc1ccc(CC#N)cc1
IUPAC-namn 2-[4-(3-chloro-2-hydroxypropoxy)phenyl]acetonitrile
Molekylvikt (g/mol) 225.67
Kvantitet 100 mg
Formel vikt 225.0557 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.