missing translation for 'onlineSavingsMsg'
Learn More

[2-[4-[(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]acetyl Chloride, TRC

Product Code. 30354971
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30354971 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354971 Supplier Toronto Research Chemicals Supplier No. TRCC42227025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material [2-[4-[(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]acetyl Chloride
CAS 776284-33-4
Molekylformel C21H24Cl2N2O2
Kvantitet 25 mg
InChI formel InChI=1S/C21H24Cl2N2O2/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)25-12-10-24(11-13-25)14-15-27-16-20(23)26/h1-9,21H,10-16H2
LEDER ClC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
IUPAC-namn 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetyl chloride
Molekylvikt (g/mol) 407.33
Formel vikt 406.1215
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.