Läs mer
2-(4-(4-Nitrophenyl)butan-2-ylamino)-1-(4-methoxyphenyl)ethanol Hydrochloride, TRC
Produktkod. 30471216
Change view
Kvantitet:
5 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471216 250 mg
30482266 5 mg
30467256 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30471216 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN503070250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1346600-92-7
Molekylformel C19H25ClN2O4
Molekylvikt (g/mol) 380.87
IUPAC-namn 1-(4-methoxyphenyl)-2-((4-(4-nitrophenyl)butan-2-yl)amino)ethan-1-ol hydrochloride
LEDER CC(CCC1=CC=C(C=C1)[N+](=O)[O-])NCC(C2=CC=C(C=C2)OC)O.Cl
Specifikationer
Kemiskt namn eller material 2-(4-(4-Nitrophenyl)butan-2-ylamino)-1-(4-methoxyphenyl)ethanol Hydrochloride
Kvantitet 250 mg
InChI formel InChI=1S/C19H24N2O4.ClH/c1-14(3-4-15-5-9-17(10-6-15)21(23)24)20-13-19(22)16-7-11-18(25-2)12-8-16;/h5-12,14,19-20,22H,3-4,13H2,1-2H3;1H
Formel vikt 380.15