Läs mer
2,4,5,6-Tetrachlorophenol-13C6, TRC
Produktkod. 18071720
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18071720 1 mg
18010320 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18071720 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2913821MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246820-81-4
Molekylformel 13C6 H2 Cl4 O
Molekylvikt (g/mol) 237.847
Synonym 2,3,4,6-Tetrachlorophenol 13C6, 2,3,4,6-Tetrachlorophenol-1,2,3,4,5,6-13C6
IUPAC-namn 2,3,4,6-tetrachloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-ol
LEDER O[13c]1[13c](Cl)[13cH][13c](Cl)[13c](Cl)[13c]1Cl
Specifikationer
Kemiskt namn eller material 2,3,4,6-Tetrachlorophenol 13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C6H2Cl4O/c7-2-1-3(8)6(11)5(10)4(2)9/h1,11H/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 235.9061
Procent renhet >95%