Läs mer
2-(4,5-Dihydro-1h-imidazol-2-yl)pyridine, TRC
Produktkod. 30365588
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365588 500 mg
30355043 250 mg
30353791 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30365588 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD177851500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7471-05-8
Molekylformel C8H9N3
Molekylvikt (g/mol) 147.18
Synonym 2-(4,5-Dihydro-1H-imidazol-2-yl)pyridine, 2-(2-Pyridinyl)-2-imidazoline, 2-(2-Pyridyl)imidazoline, 2-(2'-Pyridyl)imidazoline, 2-(Pyridin-2-yl)imidazoline, 4,5-Dihydro-2-(pyridin-2-yl)-1H-imidazole, NSC 403538
IUPAC-namn 2-(4,5-dihydro-1H-imidazol-2-yl)pyridine
LEDER C1CN=C(N1)c2ccccn2
Specifikationer
Kemiskt namn eller material 2-(4,5-Dihydro-1h-imidazol-2-yl)pyridine
Kvantitet 500 mg
InChI formel InChI=1S/C8H9N3/c1-2-4-9-7(3-1)8-10-5-6-11-8/h1-4H,5-6H2,(H,10,11)
Formel vikt 147.0796