Läs mer
2,4,5-Tribromoiodobenzene, TRC
Produktkod. 30450827
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450827 250 mg
30450602 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30450827 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT366490250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,4,5-Tribromoiodobenzene
CAS 366496-32-4
Molekylformel C6H2Br3I
Kvantitet 250 mg
Synonym 1,2,4-Tribromo-5-iodobenzene
InChI formel InChI=1S/C6H2Br3I/c7-3-1-5(9)6(10)2-4(3)8/h1-2H
LEDER Brc1cc(Br)c(I)cc1Br
IUPAC-namn 1,2,4-tribromo-5-iodobenzene
Molekylvikt (g/mol) 440.7
Formel vikt 437.6751