missing translation for 'onlineSavingsMsg'
Läs mer

2,4,5-Trifluorobenzaldehyde, 97%, Thermo Scientific™

Produktkod. p-7025491
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11480057 1 g 1g
11490057 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11480057 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B20986.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is a reactant in the enantioselective synthesis of (R)-selegiline, an anti-Parkinson′s drug, (S)-benzphetamine, an anti-obesity agent, and (S)-sitagliptin, an anti-diabetic drug.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 165047-24-5
Molekylformel C7H3F3O
Molekylvikt (g/mol) 160.095
MDL-nummer MFCD00061196
InChI-nyckel CYIFJRXFYSUBFW-UHFFFAOYSA-N
Synonym 2,4,5-trifluoro benzaldehyde, benzaldehyde, 2,4,5-trifluoro, benzaldehyde, 2,4,5-trifluoro-9ci, pubchem1468, pubchem4258, intermediates-zcf02238, acmc-209ds3, 2,4,5-trifluorbenzaldehyde, 2,4,5-trifluorobenzoaldehyde, ksc494i3d
PubChem CID 519278
IUPAC-namn 2,4,5-trifluorobenzaldehyde
LEDER C1=C(C(=CC(=C1F)F)F)C=O

Specifikationer

Kemiskt namn eller material 2,4,5-Trifluorobenzaldehyde
Densitet 1.408
Kokpunkt 174°C
Flampunkt 58°C (136°F)
Brytningsindex 1.482
Kvantitet 1 g
UN-nummer UN1989
Beilstein 8479631
Känslighet Air Sensitive
Löslighetsinformation Not miscible or difficult to mix in water.
Formel vikt 160.1
Procent renhet 97%
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.