missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-4812670
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
220227-21-4
C6H2ClF3O2S
230.59
STCZWXXRRTWRSK-UHFFFAOYSA-N
2,4,5-trifluorobenzene-1-sulfonyl chloride, 2,4,5-trifluorobenzenesulphonyl chloride, 2,4,5-trifluorophenyl sulfonyl chloride, 2,4,5-trifluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,5-trifluoro, pubchem5179, acmc-209fqs, ksc201o4t, chloro 2,4,5-trifluorophenyl sulfone, 2,4,5-trifluorophenylsulfonyl chloride
2776979
2,4,5-trifluorobenzenesulfonyl chloride
C1=C(C(=CC(=C1F)S(=O)(=O)Cl)F)F
This item is not returnable. View return policy
220227-21-4
C6H2ClF3O2S
230.59
STCZWXXRRTWRSK-UHFFFAOYSA-N
2,4,5-trifluorobenzene-1-sulfonyl chloride, 2,4,5-trifluorobenzenesulphonyl chloride, 2,4,5-trifluorophenyl sulfonyl chloride, 2,4,5-trifluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,5-trifluoro, pubchem5179, acmc-209fqs, ksc201o4t, chloro 2,4,5-trifluorophenyl sulfone, 2,4,5-trifluorophenylsulfonyl chloride
2776979
2,4,5-trifluorobenzenesulfonyl chloride
C1=C(C(=CC(=C1F)S(=O)(=O)Cl)F)F

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.6570g/mL
Flash Point >100°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula F3C6H2SO2Cl
Quantity 1 g
Specific Gravity 1.657
Formula Weight 230.59
Percent Purity 97%
Chemical Name or Material 2, 4, 5-Trifluorobenzenesulfonyl chloride
compliance-icons
Product Identifier
  • 2,4,5-Trifluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402-Store in a dry place.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'