missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4,5-Trifluorotoluene, 98%

Brand:  Alfa Aesar™ H63190.03

Additional Details : CAS Number : 887267-34-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449748

  • 190.90 SEK / 1g


Estimated Shipment date
from Supplier 03-09-2018
Add to basket

Specifications

Chemical Identifiers

887267-34-7
146.112
ZGEAYXBIJAYBKA-UHFFFAOYSA-N
17965610
C7H5F3
MFCD08458011
1,2,4-Trifluoro-5-methylbenzene

Specifications

2,4,5-Trifluorotoluene
Colorless through Yellow
98%
MFCD08458011
ZGEAYXBIJAYBKA-UHFFFAOYSA-N
1,2,4-trifluoro-5-methylbenzene
17965610
Liquid
887267-34-7
1g
C7H5F3
1,2,4-Trifluoro-5-methylbenzene
CC1=CC(=C(C=C1F)F)F
146.112
146.11
Videos
SDS