Läs mer
2,4,6-Tribromophenyl Acetate, TRC
Produktkod. 30451097
Change view
Kvantitet:
1 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30451097 2.5 g
30456921 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30451097 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7726252.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 607-95-4
Molekylformel C8 H5 Br3 O2
Molekylvikt (g/mol) 372.84
Synonym 2,4,6-Tribromo-phenol 1-Acetate, NSC 404079
IUPAC-namn (2,4,6-tribromophenyl) acetate
LEDER CC(=O)Oc1c(Br)cc(Br)cc1Br
Specifikationer
Kemiskt namn eller material 2,4,6-Tribromophenyl Acetate
Kvantitet 2.5 g
InChI formel InChI=1S/C8H5Br3O2/c1-4(12)13-8-6(10)2-5(9)3-7(8)11/h2-3H,1H3
Formel vikt 369.784