missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloropyrimidine-5-carboxaldehyde, 97%, Thermo Scientific™

Catalog Number 15492259
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
50270-27-4
C5HCl3N2O
211.426
MFCD02257700
KVJIRFGNHAAUNQ-UHFFFAOYSA-N
2,4,6-trichloro-5-pyrimidinecarboxaldehyde, 2,4,6-trichloro-pyrimidine-5-carbaldehyde, 2,4,6-trichloro-5-formylpyrimidine, 5-formyl-2,4,6-trichloropyrimidine, 2,4,6-trichloro-pyrimidine-5-carbal, 2,4,6-trichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2,4,6-trichloro, 5-pyrimidinecarboxaldehyde,2,4,6-trichloro, pubchem13189, 2,4,6-tri-chloropyrimidine-5-carbaldehyde
10932746
2,4,6-trichloropyrimidine-5-carbaldehyde
C(=O)C1=C(N=C(N=C1Cl)Cl)Cl
This item is not returnable. View return policy
50270-27-4
C5HCl3N2O
211.426
MFCD02257700
KVJIRFGNHAAUNQ-UHFFFAOYSA-N
2,4,6-trichloro-5-pyrimidinecarboxaldehyde, 2,4,6-trichloro-pyrimidine-5-carbaldehyde, 2,4,6-trichloro-5-formylpyrimidine, 5-formyl-2,4,6-trichloropyrimidine, 2,4,6-trichloro-pyrimidine-5-carbal, 2,4,6-trichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2,4,6-trichloro, 5-pyrimidinecarboxaldehyde,2,4,6-trichloro, pubchem13189, 2,4,6-tri-chloropyrimidine-5-carbaldehyde
10932746
2,4,6-trichloropyrimidine-5-carbaldehyde
C(=O)C1=C(N=C(N=C1Cl)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate. It is a versatile intermediate in the synthesis of reactive dyes.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, well ventilated area. Store cold 2-4°C.
TRUSTED_SUSTAINABILITY
Melting Point 131°C to 132°C
Color White to Brown
Quantity 1 g
UN Number UN2811
Beilstein 744505
Solubility Information Slightly soluble in water.
Formula Weight 211.43
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,4,6-Trichloropyrimidine-5-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'