missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzene-1,3,5-tricarbonitrile, TRC

Product Code. 30455751
Change view
Click to view available options
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30455751 50 mg
30455008 10 mg
30451150 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455751 Supplier Toronto Research Chemicals Supplier No. TRCT89991050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3638-97-9
Molekylformel C9N3F3
Molekylvikt (g/mol) 207.11
IUPAC-namn 2,4,6-trifluorobenzene-1,3,5-tricarbonitrile
LEDER Fc1c(C#N)c(F)c(C#N)c(F)c1C#N

Specifications

Kemiskt namn eller material 2,4,6-Trifluorobenzene-1,3,5-tricarbonitrile
Kvantitet 50 mg
InChI formel InChI=1S/C9F3N3/c10-7-4(1-13)8(11)6(3-15)9(12)5(7)2-14
Formel vikt 207.0044
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.