Läs mer
2,4,6-Trihydroxybenzoic Acid, TRC
Produktkod. 30454383
Change view
Kvantitet:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454383 100 g
30449586 50 g
30456682 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30454383 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT795065100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83-30-7
Molekylformel C7 H6 O5
Molekylvikt (g/mol) 170.12
Synonym 2,4,6-Trihydroxybenzenecarboxylic Acid, NSC 36720, Phloroglucinic Acid, Phloroglucinolcarboxylic Acid
IUPAC-namn 2,4,6-trihydroxybenzoic acid
LEDER OC(=O)c1c(O)cc(O)cc1O
Specifikationer
Kemiskt namn eller material Phloroglucinol carboxylic acid
Kvantitet 100 g
InChI formel InChI=1S/C7H6O5/c8-3-1-4(9)6(7(11)12)5(10)2-3/h1-2,8-10H,(H,11,12)
Formel vikt 170.0215