missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonyl azide, 97%, contains 15% water as stabilizer, Thermo Scientific Chemicals

Catalog Number 15239726
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
36982-84-0
C15H23N3O2S
309.43
AEMWUHCKKDPRSK-UHFFFAOYSA-N
2,4,6-triisopropylbenzenesulfonyl azide, 2,4,6-triisopropylbenzene-sulfonyl azide, trisyl azide, 2,4,6-triisopropylbenzene-sulfonyl azide,, benzenesulfonyl azide, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropyl-benzenesulfonyl azide, n-diazo-2,4,6-triisopropyl-benzenesulfonamide, 2,4,6-tris 1-methylethyl-benzenesulfonyl azide, 2,4,6-triisopropylbenzenesulfonyl azide, stab. with ca water, trisyl azide solution
371707
N-diazo-2,4,6-tri(propan-2-yl)benzenesulfonamide
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N=[N+]=[N-])C(C)C
This item is not returnable. View return policy
36982-84-0
C15H23N3O2S
309.43
AEMWUHCKKDPRSK-UHFFFAOYSA-N
2,4,6-triisopropylbenzenesulfonyl azide, 2,4,6-triisopropylbenzene-sulfonyl azide, trisyl azide, 2,4,6-triisopropylbenzene-sulfonyl azide,, benzenesulfonyl azide, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropyl-benzenesulfonyl azide, n-diazo-2,4,6-triisopropyl-benzenesulfonamide, 2,4,6-tris 1-methylethyl-benzenesulfonyl azide, 2,4,6-triisopropylbenzenesulfonyl azide, stab. with ca water, trisyl azide solution
371707
N-diazo-2,4,6-tri(propan-2-yl)benzenesulfonamide
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N=[N+]=[N-])C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 41.0°C to 43.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 309.43
Percent Purity 97%
Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl azide
compliance-icons
Product Identifier
  • 2,4,6-Triisopropylbenzenesulfonyl azide, contains 15% water as stabilizer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'