missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Triisopropylbenzoic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
2,4,6-Triisopropylbenzoic acid is a hindered acid intermediate compound in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4,6-Triisopropylbenzoic acid is a hindered acid intermediate compound in organic synthesis.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
2,4,6-Triisopropylbenzoic acid is a hindered acid intermediate compound in organic synthesis.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 184°C to 187°C |
Quantity | 10 g |
Beilstein | 2372331 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 248.37 |
Percent Purity | 97% |
Chemical Name or Material | 2,4,6-Triisopropylbenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'