missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals
512.81 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
250 g
Description
2,4,6-Trimethoxybenzaldehyde was used in the preparation and characterization of three RNA-specific fluorescent probes and their use in live cell imaging. It was used as starting reagent for the regioselective synthesis of new (±)-8-alkyl-5,7-dihydroxy-4-(4-hydroxyphenyl)-3,4-dihydrocoumarins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4,6-Trimethoxybenzaldehyde was used in the preparation and characterization of three RNA-specific fluorescent probes and their use in live cell imaging. It was used as starting reagent for the regioselective synthesis of new (+/-)-8-alkyl-5,7-dihydroxy-4-(4-hydroxyphenyl)-3,4-dihydrocoumarins.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
2,4,6-Trimethoxybenzaldehyde was used in the preparation and characterization of three RNA-specific fluorescent probes and their use in live cell imaging. It was used as starting reagent for the regioselective synthesis of new (+/-)-8-alkyl-5,7-dihydroxy-4-(4-hydroxyphenyl)-3,4-dihydrocoumarins.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | 119°C to 123°C |
Quantity | 10 g |
Beilstein | 1956051 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 196.2 |
Percent Purity | 98% |
Chemical Name or Material | 2,4,6-Trimethoxybenzaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'