Läs mer
2,4,6-trimethyl-5-pyrimidinol, TRC
Produktkod. 30329217
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30329217 100 mg
1 options
Product Code. 30329217 Supplier Toronto Research Chemicals Supplier No. TRCB587545100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 2,4,6-trimethyl-5-pyrimidinol
CAS 71267-12-4
Molekylformel C7H10N2O
Kvantitet 100 mg
InChI formel InChI=1S/C7H10N2O/c1-4-7(10)5(2)9-6(3)8-4/h10H,1-3H3
LEDER Cc1nc(C)c(O)c(C)n1
IUPAC-namn 2,4,6-trimethylpyrimidin-5-ol
Molekylvikt (g/mol) 138.17
Formel vikt 138.0793