Läs mer
2,4,6-Trimethylbenzophenone, Mikromol™
Produktkod. 17914086
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17914086 100 mg
17910956 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17914086 Leverantör Mikromol Leverantörsnummer MM2548.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2,4,6-Trimethylbenzophenone
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,4,6-Trimethylbenzophenone
CAS 954-16-5
Analyt- eller komponentnamn 2,4,6-Trimethylbenzophenone
Frakt skick Room Temperature
Molekylformel C16 H16 O
Certifikat och efterlevnad ISO 17025
Synonym Methanone, phenyl(2,4,6-trimethylphenyl)-, Benzophenone, 2,4,6-trimethyl- (6CI,7CI,8CI), 2,4,6-Trimethylbenzophenone, Benzoylmesitylene, Mesityl phenyl ketone, NSC 26923, Phenyl 2,4,6-trimethylphenyl ketone, Phenyl(2,4,6-trimethylphenyl)methanone
InChI formel InChI=1S/C16H16O/c1-11-9-12(2)15(13(3)10-11)16(17)14-7-5-4-6-8-14/h4-10H,1-3H3
LEDER Cc1cc(C)c(C(=O)c2ccccc2)c(C)c1
IUPAC-namn phenyl-(2,4,6-trimethylphenyl)methanone
Molekylvikt (g/mol) 224.3
Kvantitet 100 mg
Formel vikt 224.1201 g/mol