Läs mer
2,4,6-Trimethylbenzophenone, TRC
Produktkod. 30414773
Change view
Kvantitet:
1 g
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30414773 1 g -
30415691 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30414773 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2194051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,4,6-Trimethylbenzophenone
CAS 954-16-5
Molekylformel C16 H16 O
Kvantitet 1 g
Synonym Methanone','phenyl(2,4,6-trimethylphenyl)-, Benzophenone','2,4,6-trimethyl- (6CI,7CI,8CI), 2,4,6-Trimethylbenzophenone, Benzoylmesitylene, Mesityl phenyl ketone, NSC 26923, Phenyl 2,4,6-trimethylphenyl ketone, Phenyl(2,4,6-trimethylphenyl)methanone
InChI formel InChI=1S/C16H16O/c1-11-9-12(2)15(13(3)10-11)16(17)14-7-5-4-6-8-14/h4-10H,1-3H3
LEDER Cc1cc(C)c(C(=O)c2ccccc2)c(C)c1
IUPAC-namn phenyl-(2,4,6-trimethylphenyl)methanone
Molekylvikt (g/mol) 224.3
Formel vikt 224.1201