missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%, Thermo Scientific Chemicals

Catalog Number p-7052535
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
42333-76-6
C18H12N6
312.34
MFCD23379843
UOSYDSDIRAXTTF-UHFFFAOYSA-N
2,4,6-tris 3-pyridyl-s-triazine, 2,4,6-tri-3-pyridinyl-1,3,5-triazine, tris pyridin-3-yl-1,3,5-triazine, 2,4,6-tris 3-pyridyl-1,3,5-triazine
11472427
2,4,6-tripyridin-3-yl-1,3,5-triazine
C1=CC(=CN=C1)C1=NC(=NC(=N1)C1=CN=CC=C1)C1=CN=CC=C1
This item is not returnable. View return policy
42333-76-6
C18H12N6
312.34
MFCD23379843
UOSYDSDIRAXTTF-UHFFFAOYSA-N
2,4,6-tris 3-pyridyl-s-triazine, 2,4,6-tri-3-pyridinyl-1,3,5-triazine, tris pyridin-3-yl-1,3,5-triazine, 2,4,6-tris 3-pyridyl-1,3,5-triazine
11472427
2,4,6-tripyridin-3-yl-1,3,5-triazine
C1=CC(=CN=C1)C1=NC(=NC(=N1)C1=CN=CC=C1)C1=CN=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Color White to Yellow
Quantity 1 g
Formula Weight 312.33
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,4,6-Tris(3-pyridyl)-1,3,5-triazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'