Läs mer
2-(4-Amino-1-oxoisoindolin-2-yl)-4-carbamoyl Butyric Acid, TRC
Produktkod. 30320339
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30320339 250 mg
1 options
Produktkod. 30320339 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA619155250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Amino-2-(4-amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-5-oxopentanoic Acid
CAS 2197420-75-8
Molekylformel C13 H15 N3 O4
Kvantitet 250 mg
Synonym 2-(4-Amino-1-oxoisoindolin-2-yl)-4-carbamoyl Butyric Acid, 5-Amino-2-(4-amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-5-oxopentanoic Acid, 5-Amino-2-(4-amino-1-oxo-isoindolin-2-yl)-5-oxo-pentanoic Acid
InChI formel InChI=1S/C13H15N3O4/c14-9-3-1-2-7-8(9)6-16(12(7)18)10(13(19)20)4-5-11(15)17/h1-3,10H,4-6,14H2,(H2,15,17)(H,19,20)
LEDER NC(=O)CCC(N1Cc2c(N)cccc2C1=O)C(=O)O
IUPAC-namn 5-amino-2-(7-amino-3-oxo-1H-isoindol-2-yl)-5-oxopentanoic acid
Molekylvikt (g/mol) 277.276
Formel vikt 277.106