Läs mer
2-(4-Amino-1-oxoisoindolin-2-yl)pentanediamide, TRC
Produktkod. 30326224
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30326224 25 mg
30327816 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30326224 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61916825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(4-Amino-1-oxoisoindolin-2-yl)pentanediamide
CAS 2197414-55-2
Molekylformel C13H16N4O3
Kvantitet 25 mg
Synonym 2-(4-Amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)pentanediamide, Pentanediamide, 2-(4-amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)- (ACI), 2-(4-Amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)pentanediamide, Pentanediamide, 2-(4-amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)- (ACI)
InChI formel InChI=1S/C13H16N4O3/c14-9-3-1-2-7-8(9)6-17(13(7)20)10(12(16)19)4-5-11(15)18/h1-3,10H,4-6,14H2,(H2,15,18)(H2,16,19)
LEDER NC(=O)CCC(N1Cc2c(N)cccc2C1=O)C(=O)N
IUPAC-namn 2-(7-amino-3-oxo-1H-isoindol-2-yl)pentanediamide
Molekylvikt (g/mol) 276.3
Formel vikt 276.1222