Läs mer
2-(4-Aminophenyl)-1,2-dihydro-1-oxo-7-(2-pyridinylmethoxy)-4-(3,4,5-trimethoxyphenyl)-3-isoquinolinecarboxylic Acid Methyl Ester Dihydrochloride (>90%), TRC
Produktkod. 30327518
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30327518 50 mg
1 options
Produktkod. 30327518 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA62263550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(4-Aminophenyl)-1,2-dihydro-1-oxo-7-(2-pyridinylmethoxy)-4-(3,4,5-trimethoxyphenyl)-3-isoquinolinecarboxylic Acid Methyl Ester Dihydrochloride
CAS 212498-37-8
Molekylformel C32 H29 N3 O7 . 2 Cl H
Kvantitet 50 mg
Synonym 2-(4-Aminophenyl)-1,2-dihydro-1-oxo-7-(2-pyridinylmethoxy)-4-(3,4,5-trimethoxyphenyl)-3-isoquinolinecarboxylic Acid Methyl Ester Hydrochloride (1:2), T1032
InChI formel InChI=1S/C32H29N3O7.2ClH/c1-38-26-15-19(16-27(39-2)30(26)40-3)28-24-13-12-23(42-18-21-7-5-6-14-34-21)17-25(24)31(36)35(29(28)32(37)41-4)22-10-8-20(33)9-11-22;;/h5-17H,18,33H2,1-4H3;2*1H
LEDER Cl.Cl.COC(=O)C1=C(c2cc(OC)c(OC)c(OC)c2)c3ccc(OCc4ccccn4)cc3C(=O)N1c5ccc(N)cc5
IUPAC-namn methyl 2-(4-aminophenyl)-1-oxo-7-(pyridin-2-ylmethoxy)-4-(3,4,5-trimethoxyphenyl)isoquinoline-3-carboxylate;dihydrochloride
Molekylvikt (g/mol) 640.51
Formel vikt 639.154