Läs mer
[2-[4-(Aminosulfonyl)phenyl]ethyl]carbamic Acid Ethyl Ester, TRC
Produktkod. 30310560
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30310560 100 mg
1 options
Produktkod. 30310560 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA629490100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl [2-(4-Sulphamoylphenyl)ethyl]carbamate
CAS 192118-08-4
Molekylformel C11 H16 N2 O4 S
Kvantitet 100 mg
Synonym Carbamic acid, [2-[4-(aminosulfonyl)phenyl]ethyl]-, ethyl ester (9CI), Ethyl [2-(4-Sulphamoylphenyl)ethyl]carbamate, Ethyl [2-(4-Sulfamoylphenyl)ethyl]carbamate, [2-[4-(Aminosulfonyl)phenyl]ethyl]carbamic Acid Ethyl Ester
InChI formel InChI=1S/C11H16N2O4S/c1-2-17-11(14)13-8-7-9-3-5-10(6-4-9)18(12,15)16/h3-6H,2,7-8H2,1H3,(H,13,14)(H2,12,15,16)
LEDER CCOC(=O)NCCc1ccc(cc1)S(=O)(=O)N
IUPAC-namn ethyl N-[2-(4-sulfamoylphenyl)ethyl]carbamate
Molekylvikt (g/mol) 272.32
Formel vikt 272.0831