Läs mer
2,4-Bis(1-methylethyl)phenyl Diphenyl Ester Phosphoric Acid, TRC
Produktkod. 30337585
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30337585 50 mg
1 options
Produktkod. 30337585 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB45148050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,4-Bis(1-methylethyl)phenyl Diphenyl Ester Phosphoric Acid
CAS 96107-55-0
Anteckningar om renhetsgrad HPLC
Molekylformel C24 H27 O4 P
Kvantitet 50 mg
Synonym 2,4-diisopropylphenyl Diphenyl Phosphate
InChI formel InChI=1S/C24H27O4P/c1-18(2)20-15-16-24(23(17-20)19(3)4)28-29(25,26-21-11-7-5-8-12-21)27-22-13-9-6-10-14-22/h5-19H,1-4H3
LEDER CC(C)c1ccc(OP(=O)(Oc2ccccc2)Oc3ccccc3)c(c1)C(C)C
IUPAC-namn (2,4-diisopropylphenyl) diphenyl phosphate
Molekylvikt (g/mol) 410.44
Formel vikt 410.1647