missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Bis-(trifluoromethyl)pyrimidine-5-carboxylic acid ethyl ester, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 188781-15-9 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C9H6F6N2O2 |
Quantity | 1 g |
Synonym | ethyl 2,4-bis trifluoromethyl pyrimidine-5-carboxylate, 2,4-bis-trifluoromethyl pyrimidine-5-carboxylic acid ethyl ester, 2,4-bis-trifluoromethyl pyrimidine-5-carboxylicacid ethyl ester, 2,4-bis trifluoromethyl-5-ethoxycarbonyl pyrimidine, ethyl-2,4-bis trifluoromethyl pyrimidine-5-carboxylate, ethyl 2,4-bis-trifluoromethyl pyrimidine-5-carboxylate, 2,4-bis-trifluoromethylpyrimidine-5-carboxylic acid ethyl ester, 2,4-bis-trifluoromethyl-pyrimidine-5-carboxylic acid ethyl ester, ethyl 2-trifluoromethyl-4-trifluoromethyl-5-pyrimidine carboxylate |
InChI Key | SRFVAGSJHJMMME-UHFFFAOYSA-N |
SMILES | CCOC(=O)C1=CN=C(N=C1C(F)(F)F)C(F)(F)F |
IUPAC Name | ethyl 2,4-bis(trifluoromethyl)pyrimidine-5-carboxylate |
Molecular Weight (g/mol) | 288.149 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'