missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Bromo-2-thienyl)-5-methylbenzimidazole, 95%

Brand:  Alfa Aesar™ H64532.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15471789

  • 2834.60 SEK / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Bromo-2-thienyl)-5-methylbenzimidazole
293.18
MFCD26967924

Specifications

2-(4-Bromo-2-thienyl)-5-methylbenzimidazole
95%
MFCD26967924
Solid
293.18
1g
Videos
SDS