missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Bromophenyl)-2-propanol, 97%, Thermo Scientific Chemicals
1554.24 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a medicine intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a medicine intermediate.
Solubility
Slightly soluble in water (1.4 g/L) (25°C)
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a medicine intermediate.
Solubility
Slightly soluble in water (1.4 g/L) (25°C)
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Melting Point | 40°C to 45°C |
Color | White |
Density | 1.356 g/mL |
Boiling Point | 75°C to 85°C (0.1 mmHg) |
Refractive Index | 1.552 |
Quantity | 1 g |
Beilstein | 1935066 |
Solubility Information | Slightly soluble in water (1.4g/L) (25°C) |
Formula Weight | 215.09 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'