missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Bromophenyl)thiazolidine, 97%

Brand:  Alfa Aesar™ H62698.03

Additional Details : CAS Number : 67086-81-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489178

  • 2477.86 SEK / 1g


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Bromophenyl)thiazolidine
67086-81-1
244.15
MFCD00985113

Specifications

2-(4-Bromophenyl)thiazolidine
1g
244.15
67086-81-1
97%
MFCD00985113
Videos
SDS