Läs mer
2-[(4-Chloro-2-nitrophenyl)amino]benzoic Acid, TRC
Produktkod. 30368351
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368351 250 mg
30364905 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30368351 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC373950250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 60091-87-4
Molekylformel C13H9ClN2O4
Molekylvikt (g/mol) 292.67
Synonym Clozapine Impurity 7
IUPAC-namn 2-(4-chloro-2-nitroanilino)benzoic acid
LEDER OC(=O)c1ccccc1Nc2ccc(Cl)cc2[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 2-[(4-Chloro-2-nitrophenyl)amino]benzoic Acid
Kvantitet 250 mg
InChI formel InChI=1S/C13H9ClN2O4/c14-8-5-6-11(12(7-8)16(19)20)15-10-4-2-1-3-9(10)13(17)18/h1-7,15H,(H,17,18)
Formel vikt 292.0251