Läs mer
2-(4-Chloro-3-sulfobenzoyl)-benzoic Acid, TRC
Produktkod. 30133231
Change view
Kvantitet:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133231 250 mg
30135170 100 mg
30009057 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133231 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC427810250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 345930-32-7
Molekylformel C14 H9 Cl O6 S
Molekylvikt (g/mol) 340.74
Synonym Benzoic acid, 2-(4-chloro-3-sulfobenzoyl)-, 2-(4-Chloro-3-sulfobenzoyl)benzoic acid
IUPAC-namn 2-(4-chloro-3-sulfobenzoyl)benzoic acid
LEDER OC(=O)c1ccccc1C(=O)c2ccc(Cl)c(c2)S(=O)(=O)O
Specifikationer
Kemiskt namn eller material 2-(4-Chloro-3-sulfobenzoyl)-benzoic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C14H9ClO6S/c15-11-6-5-8(7-12(11)22(19,20)21)13(16)9-3-1-2-4-10(9)14(17)18/h1-7H,(H,17,18)(H,19,20,21)
Formel vikt 339.9808
Procent renhet >95%